Carminic acid
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC11044000EA
927
DKK
EHERC11044000
Carminic acid
Carminic acid
Formel:
C₂₂H₂₀O₁₃ |
MDL Number:
MFCD00167028 CAS-nummer: 1260-17-9 EINECS: 215-023-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...