Electrode holder, flexible, SH300
Leverandør: Consort
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CONSSH300EA 2000 DKK
CONSSH300
Electrode holder, flexible, SH300
Model SH300 holds up to three standard electrodes. It's heavy base and very stable flexible arm allow the electrodes to move sideways or up and down while keeping them at a constant vertical angle.