Spiral plater, WASP 2
Leverandør: Don Whitley Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WASP 2
710-0915EA 114790 DKK
710-0915
Spiral plater, WASP 2
Spiralformede udpladere
Spiral platers are used extensively in the food industry to test for safe levels of bacteria; non-food applications include pharmaceuticals, cosmetics, water, and mouth and skin bacteria. The WASP 2 (Whitley Automatic Spiral Plater) takes advantage of contemporary technology to offer microprocessor-controlled stepper motor motion to streamline and automate traditional spiral plate techniques.
  • Eliminates the need for serial dilutions
  • Requires minimal set up time
  • Reduces cost per test
  • Standardises counting methods
Leveringsinformation: Includes water pots, spare bleach bottle, disposable beakers and tool kit.