Culture media preparator, MasterClave 09
Leverandør: bioMérieux
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MasterClave®
710-0702EA 197620 DKK
710-0702
Culture media preparator, MasterClave 09
Mediefremstillingsapparat
The Masterclave® 09 is an easy-to-operate, automated culture media preparator for standardised culture media preparation. The Masterclave® 09 can be connected to Petri dish filling machines and gravimetric sample diluters. It offers a high level of flexibility and several safety features. The alphanumeric keyboard can be used to save up to 40 programs. Features a safety lid with transparent polycarbonate window, secured by means of a magnetic lock. The patented stainless steel magnetic stirrer ensures consistent results.
  • Capacity: 1 to 9 litres
  • Full documentation
  • Very easy to operate and clean, mobile
  • Up to 40 user-specific programs
  • Full cycle lasts just 1 hour and 15 minutes