5-Chlor-2-nitrobenzoesyre 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
109680100
109681000
ACRO109680100EA
154
DKK
ACRO109680100
ACRO109681000
5-Chlor-2-nitrobenzoesyre 99%
5-Chlor-2-nitrobenzoesyre
Formel:
C₇H₄ClNO₄ Smeltepunkt: 136…139 °C Massefylde: 1,593 g/cm³ (18 °C) Flammepunkt: >100 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007290 CAS-nummer: 2516-95-2 EINECS: 219-738-1 |
Specifikationer Testresultater
Appearance | Light yellow to yellow-green Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 136 to 139 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥98.5 % |
Nickel (Ni) | ≤50 ppm |
Zinc (Zn) | ≤200 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...