Vials med bred hals til skruelåg 9 mm, Chromacol™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Chromacol™
548-1560EA 379 DKK
548-1560 548-1542 548-1211
Vials med bred hals til skruelåg 9 mm, Chromacol™
Hætteglas
These high performance wide opening SureStop™ screw thread vials are made from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass, and meet all US, EU and JPN Pharmacopia standards.
  • Consistent seal, independent of user applied torque, when closing the vial
  • Optimum septum compression across the opening of the vial, irrespective of torque applied
  • Designed to be consistently flat across the top making it more compatabile for autosamplers
SureStop™ 9 mm vials, are designed as part of the Advance Vial Closure System (AVCS), and remove any subjectivity around achieving the optimal seal compression when closing a vial. As an integral component of AVCS technology, SureStop™ vials offer the sealing and performance characteristics of a crimp top vial and the versatility and convenience of a threaded vial. This is achieved by incorporating a definite stop point into the design of the vial finish, preventing over tightening of the closure. SureStop™ vials remove all doubt about the degree of tightening necessary for optimal sealing.
Certificeringer: Meets ASTM standards. EN-342.
Leveringsinformation: The convenience and instrument select kits include 100 vials and 100 caps with pre-assembled septa for added convenience during sample preparation.
Emballering: Packed in reusable two compartment trays to keep matching supplies together.