Cell culture bottles, Breed-Demeter, square, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
391-0690EA 227 DKK
391-0690
Cell culture bottles, Breed-Demeter, square, DURAN®
Flasker Medieflasker
DURAN®, borosilikatglas, 3.3, klar.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For cell and tissue culture.