VWR® Hichrom Last Drop™ Inlet Filters
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
554-7723EA 653 DKK
554-7723 554-7724 HICHHI-692
VWR® Hichrom Last Drop™ Inlet Filters
Filtre Væskebehandlingsfiltre
The Last Drop™ eluent filter utilises a flat element, which sits parallel to the bottom of the reservoir, allowing the filter to draw all but the last 2% of the eluent without drawing air into the system. The filter consists of a 316 stainless steel or PTFE filter element in an inert housing.
  • Reduces solvent waste
  • Should be used for sensitive biochromatography applications
Leveringsinformation: Every filter is supplied with a universal PEEK tubing adaptor.