VWR® Hichrom Pliers, for Stainless Steel Tubing
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Avantor® Hichrom
554-7744EA 550 DKK
554-7744
VWR® Hichrom Pliers, for Stainless Steel Tubing
Tænger
These pliers are designed for cutting 1/16" stainless steel tubing and are especially suitable for tubing of 0,010" and 0,020" ID.
  • Leaves smooth and uniform cuts
  • De-burring may be necessary to achieve an optimum finish