VWR®, Podeøser og podenåle i dispenserrør
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
612-9359EA 640 DKK
612-9359 612-9366 612-9362 612-9361 612-9360
VWR®, Podeøser og podenåle i dispenserrør
Spredere Podenåle og Loops
PS, steril.
  • Colour coded for easy product identification
  • Hexagonal loop shaft improves grip, assisting orientation and making diluting and streaking easier
  • Gamma sterilised
The resealable pouch allows loops to be easily accessed any number of times, then re-closed to maintain product integrity. The loops and needles are positioned upside down in the pouch in order to reduce any risk of contamination. Optional pack holder facilitates easy access using one hand, leaving the other hand free to hold a culture plate or tube.
Leveringsinformation: Leveres med et kalibreringscertifikat i hver kasse.
Emballering: 40 løkker/pose (25 poser pr. pakke).