Seals for flanged connections
Leverandør: Lenz
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
201-0126EA 129 DKK
201-0126
Seals for flanged connections
Forsegling Mechanical Seals
PTFE, hvid
  • Simple easy assembly and disassembly
  • Very good chemical resistance
  • Very good thermal resistance
For impermeable ball or flat flange connections.