EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kits
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17881 17881RF
STMC17881EA 7358 DKK
STMC17881 STMC17881RF
EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kits
Celle separations medie
Immunomagnetic positive selection of human CD15+ cells (e.g. myeloid cells) from whole blood.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit is designed to isolate highly purified CD15+ cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD15 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD15 antigen is expressed on granulocytes and monocytes.
This product replaces the EasySep™ Human Whole Blood CD15 Positive Selection Kit and EasySep™ HLA Whole Blood CD15 Positive Selection Kit, for even faster cell isolations.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').