EasySep™ RBC Depletion Reagents, 10 ml
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
18170 18170RF
STMC18170EA 1730 DKK
STMC18170 STMC18170RF
EasySep™ RBC Depletion Reagents, 10 ml
Celle separations medie
Immunomagnetic isolation of untouched human leukocytes from whole blood.
  • 99,9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ RBC Depletion Reagent is designed to immunomagnetically deplete red blood cells (RBCs) from fresh human peripheral blood, umbilical cord blood, bone marrow or leukapheresis samples by negative selection. RBCs are targeted with Tetrameric Antibody Complexes recognising Glycophorin A and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube while desired cells are poured off.
This product replaces the EasySep™ Human Glycophorin A Depletion Kit for even faster cell isolations.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').