EasySep™ Rat IgM Depletion Kits
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
18644 18644RF
STMC18644EA 7711 DKK
STMC18644 STMC18644RF
EasySep™ Rat IgM Depletion Kits
Celle separations medie
Immunomagnetic depletion of rat IgM+ cells.
  • Fast, easy-to-use and column-free
  • Up to 90% purity (≤10% IgM+)
  • Isolated cells are untouched
The EasySep™ Rat IgM Depletion Kit is designed to deplete IgM+ cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by positive selection. The EasySep™ procedure involves labelling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').