EasySep™ HLA B Cell Enrichment Kits
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
19054HLARF 19054HLA
STMC19054HLARFEA 8198 DKK
STMC19054HLARF STMC19054HLA
EasySep™ HLA B Cell Enrichment Kits
Celle separations medie
Column-free immunomagnetic isolation of untouched human B cells for HLA analysis.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
  • Compatible with downstream HLA assays
The EasySep™ HLA B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-B cells and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnetMore Product Information