EasySep™ Human CD34 Positive Selection Kit II
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17856 17856RF
STMC17856EA 7820 DKK
STMC17856 STMC17856RF
EasySep™ Human CD34 Positive Selection Kit II
Celle separations medie
Immunomagnetic positive selection of human CD34+ cells (e.g. hematopoietic, stem/progenitor cells)
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human CD34 Positive Selection Kit II is designed to isolate CD34+ cells from fresh or previously frozen bone marrow or mobilized peripheral blood mononuclear cells, from previously frozen cord blood mononuclear cells, or from hESC/hiPSC-derived hematopoietic cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD34 and dextran-coatedMore Product Information