EasySep™ Human Pan-Granulocyte Isolation Kits
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
19259 19259RF
STMC19259EA 8039 DKK
STMC19259 STMC19259RF
EasySep™ Human Pan-Granulocyte Isolation Kits
Celle separations medie
Immunomagnetic isolation of untouched human pan-granulocytes
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human Pan-Granulocyte Enrichment Kit is designed to isolate granulocytes (eosinophils, basophils and neutrophils) from normal peripheral whole blood by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognising non-granulocytes and magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').