EasySep™ Direct Human B Cell Isolation Kits
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
19674 19674RF
STMC19674EA 5334 DKK
STMC19674 STMC19674RF
EasySep™ Direct Human B Cell Isolation Kits
Celle separations medie
Immunomagnetic isolation of B cells directly from whole blood
  • >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 95% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
Isolate highly purified B cells directly from human whole blood without the need for density gradient centrifugation, RBC sedimentation or lysis. This platform provides fast and fully immunomagnetic negative selection.
This cell isolation kit targets non-B cells for removal with EasySep™ Direct RapidSpheres™ and antibodies recognising specific surface antigens. Unwanted cells are separated from desired cells using an EasySep™ magnet, and untouched target cells are then pipetted or poured into a new tube.
Isolated cells can be used directly in downstream applications such as flow cytometry, culture or DNA/RNA extraction.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').