EasySep™ Mouse NK Cell Isolation Kits
Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
19855 19855RF
STMC19855EA 6325 DKK
STMC19855 STMC19855RF
EasySep™ Mouse NK Cell Isolation Kits
Celle separations medie
Immunomagnetic isolation of untouched mouse NK cells
  • Fast and easy-to-use
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse NK Cell Isolation Kit targets non-NK cells by labeling unwanted cells with biotinylated antibodies and EasySep™ Streptavidin RapidSpheres™, and separates cells without columns using an EasySep™ magnet. Desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays.
This product replaces the EasySep™More Product Information