HOLDEX® Single-Use Blood Culture Holder
Leverandør: Greiner bio-one
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HOLDEX®
GREI450241_800EA 0 DKK
GREI450241_800
HOLDEX® Single-Use Blood Culture Holder
Kanyleholder
The HOLDEX® Single-use holder is a sterile device with a male Luer slip connector to enable compatibility with standard female Luer connections. The excentric position of this Luer connection offers the further advantage of a comfortable, flat puncture angle.
  • Single-packed, sterile, not made with natural rubber latex
  • An innovative solution for streamlining the product line
  • One blood collection holder for different puncture devices
  • Optimal puncture angle for ease of use
  • Optical blood flow indicator directly at the puncture site
HOLDEX® was especially developed for blood collection from difficult vein conditions. Due to the Luer connector which is positioned off-centre (excentric), the user has a pleasant, flat puncture angle. This increases comfort for both user and patient.