Mouse monoclonal IgM antibody against human, chimpanzee CD66b, FITCconjugated.
- Granulocytes
- Subsets
- Myeloid cells
The G10F5 antibody reacts with CD66b (previously denoted CD67), an ~95 kDa single-chain glycosylphosphatidylinositol (GPI)-linked glycoprotein expressed by neutrophilic and eosinophilic granulocytes. CD66b is a member of the carcinoembryonic antigen (CEA)-like subfamily of proteins within the immunoglobulin superfamily and possesses two Ig-like C2-type domains and one Ig-like V-type domain. In neutrophils, CD66b is localized within granules in the cytoplasm as well as being expressed on the plasma membrane. Surface expression levels increase upon granulocyte activation, an effect induced by stimulators such as phorbol myristate acetate (PMA), calcium ionophore, and N-formylmethionyl-leucyl-phenylalanine (FMLP). CD66b appears to play a role as a signaling receptor involved in cell adhesion, phagocytosis, and chemotaxis by regulating granulocyte activation and adhesion to several proteins, including CD66c, galectin-3, fibronectin, and E-selectin.
Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Type:
Antigen: CD66b
Clonality: Monoclonal
Clone: G10F5
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Chimpanzee;Human