Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (APC)

Leverandør: STEMCELL Technologies
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

60082AZ 60082AZ.1
STMC60082AZEA 3049 DKK
STMC60082AZ STMC60082AZ.1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (APC)
Antistoffer
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD5, APC­conjugated.

  • T cells

The UCHT2 antibody reacts with CD5, an ~67 kDa single-chain type I glycoprotein and member of the scavenger receptor superfamily, which is constitutively expressed on thymocytes, T cells, B cell subsets, peripheral blood dendritic cells and some leukemia and lymphoma cells, including chronic B lymphocytic leukemia (B-CLL) cells. CD5 is expressed at low levels on thymocytes and at high density on mature T cells. Putative ligands include CD5L and CD72. CD5 modulates T and B cell receptor signaling, thymocyte maturation, and T cell/B cell interactions via its physical and functional associations with the T cell receptor/CD3 complex and the B cell receptor. Studies with CD5 knockout mice indicate that CD5 negatively regulates antigen receptor-mediated signaling in thymocytes and mature T cells. The UCHT2 antibody binds to epitope 2 located in the extracellular domain of CD5.

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (APC)
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Capuchin monkey, Chimpanzee, Common marmoset, Cynomolgus monkey, Human, Owl monkey, Rhesus monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR