Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy3®)

Leverandør: AAT Bioquest
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AATB16862EA 2210 DKK
AATB16862
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy3®)
Antistoffer
AAT Bioquest's anti-mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity-purified
  • Bright signal
  • Cross-adsorbed

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR