Native Horseradish Horseradish peroxidase (from Serum), Cinoxacin
Leverandør: AAT Bioquest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AATB50022EA 5520 DKK
AATB50022
Native Horseradish Horseradish peroxidase (from Serum), Cinoxacin
Proteiner
Cinoxacin-HRP conjugate is used for developing an ELISA assay for quantifying cinoxacin.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Advarsel: For research use only.