Z-IETD-pNA
Leverandør: AAT Bioquest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AATB13413EA 2210 DKK
AATB13413
Z-IETD-pNA
Proteiner
Z-IETD-pNA is a colorimetric caspase-8/granzyme B substrate containing the benzyloxycarbonyl (Z) moiety.
  • Hazard Class/ Packing Group: 5, II
  • GHS symbol: GHS07
  • Signal Word: Warning