Streptavidin, RPE
Leverandør: AAT Bioquest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AATB16901EA 3300 DKK
AATB16901 AATB16900
Streptavidin, RPE
Proteiner
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
  • Extremely high biotin-binding affinity
  • High quality conjugate
Advarsel: For research use only.