nanoPAC Power Supply
Leverandør: NanoEnTek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NANOPAC-300PEA 7040 DKK
NANOPAC-300P
nanoPAC Power Supply
Strømforsyninger
The nanoPAC mini power supply are an ultra compact and economic unit ideal for use with DNA (horizontal) and protein (vertical) electrophoresis systems.
  • In-built safety features
  • Conspicuous three digit LED
  • Alarm function
  • Wipe-clean polycarbonate housing
The maximum constant voltage up to 300 or 500V and maximum constant current output of 400 mA the nanoPACs are capable of running all cleaver scientific horizontal multiSUB systems and vertical mini.
The nanoPACs user- friendly interface is easily adjustable in 1V and 1 mA increments, making it perfect for separations where precise settings are required. Its ultra compact size and two pairs of parallel power terminals, which can run two electrophoresis units simultaneously, save time and bench space.
Certificeringer: ETL, FCC.
Leveringsinformation: This unit has a 12 month limited warranty.
Emballering: 1x nanoPAC 300 power supply; 1x power cord; 1x instruction manual.