VWR®, EPA Method 1633 Reference Standard, 100 µg/L in acetonitrile, CRM
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
89090.010EA 0 DKK
89090.010
VWR®, EPA Method 1633 Reference Standard, 100 µg/L in acetonitrile, CRM
Standarder Miljø analyse standarder
USEPA Method 1633 is for analysis of PFAS in aqueous, solid, biosolids and tissue samples using LC-MS/MS technique. Our mixes are offered to cover the 40 native PFAS required by the method.
Contains:
N-Methylperfluorooctanesulfonamidoethanol 24448-09-7 100 µg/mL
N-Ethyl-N-(2-hydroxyethyl)perfluorooctylsulphonamide 1691-99-2 100 µg/mL
Certificeringer: ISO 17034