Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07514-10EA 4210 DKK
MFLX07514-10 MFLX07514-30 MFLX07514-40
Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Pumper Pumpehoveder Peristaltiske pumpehoveder
Save time with fast tubing changes and reduced maintenance.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® Precision pump tubing sizes
This Easy-Load® pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the followingMore Product Information