Masterflex® L/S® Easy-Load® Head with Open-Head Sensor for Precision Tubing, 4- Roller, PARA Housing, SS Rotor
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR Item#MFLX07514-30
Your Organization's Price:
Save time with fast tubing changes and reduced maintenance.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® Precision pump tubing sizes
  • Models with open-head interlock sensor stop drive motor when pump head is open (when used with sensor-equipped drives)
This Easy-Load® pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18.
More Product Information
Other Products In this Family
See all Items in this Product Family