Tissue DNA extraction kit, Precellys®
Leverandør: Bertin
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Precellys®
BERTD05701.50EAEA 1870 DKK
BERTD05701.50EA
Tissue DNA extraction kit, Precellys®
Molekylær biologiske reagenser Nucleic Acid Purification Kits and Reagents
Precellys® homogenisers and nucleic acid extraction kits offer an optimised solution to purify DNA from your tissue samples.
  • Premium quality: High yields of purified DNA
  • Optimised Precellys® workflow from tissue to nucleic acid
  • Easy to use: Spin column technology
  • Versatile: Single kits for multiple tissue types
Our Precellys® tissue DNA extraction kit allow for the extraction and purification of DNA from a variety of tissue samples such as heart, liver, ear or tail. The silica based mini column technology delivers highly pure nucleic acids that are free from protein and other contaminants and compatible with a large range of downstream applications such as PCR, RT-qPCR, sequencing, genotyping, Southern blot and Northern analysis.
Leveringsinformation: The kits include a comprehensive user guide with optimised protocols for tissue homogenisation and nucleic acid purification.
Advarsel: Store at room temperature. For research use only.