Anaerobic systems, workstation, Whitley A95
Leverandør: Don Whitley Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Whitley
DOWIA07400EA 719150 DKK
DOWIA07400
Anaerobic systems, workstation, Whitley A95
Anaerobe systemer
The A95 Workstation has a huge capacity of up to 1120×90mm Petri dishes, depending upon the way you choose to configure your working and incubation areas.
  • Innovative touch screen interface, which allows the incorporation of some unique features and benefits
  • The four manually operated, oval glove ports enable two people to operate within the same controlled atmosphere simultaneously
  • Rapid, built-in airlock with mechanical and electrical interlocks -no risk of compromising conditions inside the chamber, flushed with nitrogen as an additional cost saving
  • Optional anaerobic conditions monitor
  • Dual gas operation for real savings on gas
  • Automatic commissioning cycle
  • Automatic de-humidification system – no user maintenance necessary
  • Optional Three Gas (TG) version available to help reduce running costs and provide additional flexibility
This model is ideal for tutoring or for very busy laboratories. As well as having a huge 36 litre airlock, each port also acts as a mini airlock so you can transfer 10 plates when you insert or withdraw each arm, reducing gas usage and the number of airlock cycles required.
Note: The cycle time mentioned in the specifications is for the airlock.
Bestillingsoplysninger: Options and accessories not included (sold separately). For more information on the available options and accessories, please see the attached brochures and contact your Avantor representative.