SARS-CoV-2 Antigen rapid test cassette (nasal swab)
Leverandør: SureScreen Diagnostics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
COVID19AGVCGEA 1570 DKK
COVID19AGVCG
SARS-CoV-2 Antigen rapid test cassette (nasal swab)
Assays Immunologiske Assays
A rapid test for the qualitative detection of SARS-CoV-2 antigen in a nasal swab.
INTENDED USE:
The SARS-CoV-2 antigen rapid test cassette is a rapid chromatographic immunoassay for the qualitative detection of SARS-CoV-2 antigen in a Nasal swab. The identification is based on monoclonal antibodies specific to the Nucleocapsid (N) protein of SARS-CoV-2. It is intended to aid in the rapid differential diagnosis of COVID-19 infection.
Advarsel: For professional in vitro diagnostic use only.