Biomass detection kit ULW10 Clean-Trace™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MMMPULW10EA 697 DKK
MMMPULW10
Biomass detection kit ULW10 Clean-Trace™
Overvågningssystem Hygiejne Monitoreringssystemer
Water test kit using ATP bioluminescence to detect overall biological load in a sample.
  • Assesses the amount of biomass in process waters, waste waters, water tanks and cooling towers by determining the total ATP present
  • Helps determine the biological status of process equipment and the effects of biocide treatments
  • Useful for testing aqueous samples requiring filtration and/or extraction such as those that are viscous or that contain particulates or fibres
Provides expanded flexibility over single shot devices, allowing sample clean up in cases of high quenching, particulates or fibres in aqueous samples. Best suited for laboratories and ideal for process water, wastewater, water tanks and cooling towers.
The test is used with the 3M™ Clean-Trace™ NG Luminometer for the assessment of the hygienic status of process waters by the measurement of total ATP.