Wipes, pre-saturated IPA Wipers
Leverandør: Texwipe
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-2569EA 2820 DKK
115-2569
Wipes, pre-saturated IPA Wipers
Papirservietter Engangsservietter og - håndklæder Imprægnerede servietter
IPA Wipers are saturated with isopropyl alcohol and are ideal for most general cleaning applications.
  • IPA Wipers contain a mixture of 70% IPA and 30% deionised water
  • Packed in an environmentally friendly, resealable zip bag
  • Easily disposable
They enable contamination-free work surfaces and are ideally suited for use throughout the production process, in non critical and non food production areas, as well as for cleaning surfaces around office spaces and other non production related areas.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904