Sodium heptanoate ≥97.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAE0009-25GEA 1240 DKK
TCIAE0009-25G
Sodium heptanoate ≥97.0% (by titrimetric analysis)
Sodium heptanoate
Formel: C₇H₁₄O₂
Molekylvægt: 152,17 g/mol
MDL Number: MFCD00070499
CAS-nummer: 10051-45-3
EINECS: 233-180-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) 97.0 to 101.0 % (after drying)
Solubility in Water almost transparency
Drying loss max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR