Gasprofi 1 SCS micro school edition
Leverandør: WLD-TEC GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WART6.007.000EA 3700 DKK
WART6.007.000
Gasprofi 1 SCS micro school edition
The safety burner gasprofi 1 micro school edition offers the highest safety standards when handling the flame. Suitable for connection to the fully automatic inoculation loop carousel autoloop Pro.
  • Safe gas burner for flaming and heating combines all functional advantages
  • Flame is automatically reignited
  • Direct switching on and off of the flame by pressing a button without any matches
  • Can also be operated using a foot pedal or an external IR motion sensor
The flame can be precisely adjusted with the dual button for gas and air adjustment. All external components are made of stainless steel and guarantee the highest degree of sterility. Spilled liquids flow out through a channel.
Ideal for connection to natural gas or propane or butane gas.