Avantor® Hichrom, Inlet Solvent Filters
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HICHHI-583EA 337 DKK
HICHHI-583 HI-584
Avantor® Hichrom, Inlet Solvent Filters
Filtre Væskebehandlingsfiltre
These solvent inlet filters are used at the low pressure inlet side of the pump to help protect check valves, injector and column from particulate contamination from the eluent.
  • Made from stainless steel of 10 μm porosity
  • Helps to remove dissolved gases
The large surface area filters out particulate matter from the solvent that may otherwise damage expensive hardware. No tools are required for replacement.