Organic reference standard, Theobromine, analytical standard, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42993-100MGEA 352 DKK
42993-100MG
Organic reference standard, Theobromine, analytical standard, Supelco®
Standarder Økologiske standarder
Theobromine is an active bronchodilator drug ingredient used in the treatment of acute and chronic asthma. It is a naturally present alkaloid in a wide variety of foods and beverages, such as coffee, cocoa, tea, yerba mate and energizing drinks.
Phosphosdiesterase inhibitor, weak adenosine receptor antagonist; diuretic, smooth muscle relaxant.
Theobromine may be used as a reference standard for the determination of theobromine metabolites in human plasma and urine after consumption of soluble cocoa products with different methylxanthine contents by high-performance liquid chromatography and diode array detection (HPLC-DAD) method.