Organic reference standard, N-Butyrylglycine, analytical standard, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
94274-250MGEA 2510 DKK
94274-250MG 94274-10MG 94274-50MG
Organic reference standard, N-Butyrylglycine, analytical standard, Supelco®
Standarder Økologiske standarder
Butyrylglycine is an acyl glycine. Acyl glycines are normally minor metabolites of fatty acids. However, the excretion of certain acyl glycines is increased in several inborn errors of metabolism. In certain cases the measurement of these metabolites in body fluids can be used to diagnose disorders associated with mitochondrial fatty acid beta-oxidation. Acyl glycines are produced through the action of glycine N-acyltransferase which is an enzyme that catalyzes the chemical reaction: acyl - CoA + glycine ↔ CoA + N-acylglycine amino acids composed of glycine substituted at the nitrogen rather than the usual carbon position, resulting in the loss of hydrogen bonding donors.