Deoxyoligo B2, BIOARRAY™
Leverandør: Enzo Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ENZOENZ42662150EA 3850 DKK
ALEXENZ-42662-30 ALEXENZ-42662-150 ENZOENZ42662150
Deoxyoligo B2, BIOARRAY™
Microarray reagenser
BIOARRAY™ Deoxyoligo B2 is a synthetic biotin-labelled sequence that can be used in eukaryotic expression assays and in aiding software programs by aligning the grid with the array image.
The control Deoxyoligo B2, together with the eukaryotic hybridization controls (included in Cat. No. ENZ-42661-30 or ENZ-42661-150), provides an internal control for assaying quality and performance. This is done by adding the controls to biotin-labeled target cRNA before hybridisation to probe sequences on the array.
Leveringsinformation: Deoxyoligo B2 is supplied as a 3 nM solution.