Sterilisation indicator tape
Leverandør: Megro
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MEGR340004EA 309 DKK
MEGR340004
Sterilisation indicator tape
Sterilisationsindikatorer
Adhesive tape for sterilisation with hot-air indicator.
  • Process indicator for the hot air sterilisation
  • Recommended for the use of fabrics for single use mostly for non-woven fabric
  • After the sterilisation the colour of the indicator will change into dark brown
Temperature range: recommended 160 to 180 °C / 40 min.
Thickness: 0,14 mm.