Anti-POLY Mouse Monoclonal Antibody [clone: 10H]

Leverandør: ProSci
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PRSI36-169EA 4360 DKK
PRSI36-169
Anti-POLY Mouse Monoclonal Antibody [clone: 10H]
Antistoffer
Processes such as transcription, repair and replication that require efficient DNA recognition are dependent on modulation of chromatin structure. Chromatin relaxation is a critical event that occurs during DNA repair and is associated with the negatively charged polymer of adenosine 5'-diphosphate (ADP)-ribose (PAR). PAR is synthesized from nicotinamide adenine dinucleotide (NAD+) by the poly(ADP-ribose) polymerase protein family (PARPs), of which PARP-1 (and to a lesser extent PARP-2) respond to DNA-strand breaks. PARP-1 is selectively activated by DNA strand breaks to catalyze the addition of long branched chains of PAR to a variety of nuclear proteins, most notably PARP itself. The amount of PAR formed in living cells with DNA damage is commensurate with the extent of the damage. Under DNA damage conditions, PAR undergoes a rapid turnover, with a half-life in the range of minutes, as PAR is rapidly hydrolyzed and converted to free ADP-ribose by the enzyme poly(ADP-ribose)glycohydrolase (PARG). PAR regulates not only cell survival and cell-death programmes, but also an increasing number of other biological functions with which novel members of the PARP family have been associated. These include transcriptional regulation, cell division, intracellular trafficking, inflammation and energy metabolism.

Flow Cytometry: (see A. Kunzmann, et al.; Immun. Ageing 3, 8 (2006)). Immunocytochemistry: (5-20ug/ml). Immunohistochemistry (paraffin sections): Dilute in 5% non fat dried milk in PBS to a final concentration of 5-20ug/ml). Western Blot: Incubate 2.5ug/ml in PBS, 0.05% Tween20, 5% non fat dried milk. Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: POLY
Clonality: Monoclonal
Clone: 10H
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Drosophila, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR