Natriumchlorid concentrate for dilution to 0.1 mol/l i vandigopløsning, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4684.EA 204 DKK
4684.
Natriumchlorid concentrate for dilution to 0.1 mol/l i vandigopløsning, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium chloride
Formel: NaCl
Molekylvægt: 58,44 g/mol
MDL Number: MFCD00003477
CAS-nummer: 7647-14-5
Bestil nu
Specifikationer Testresultater
For Laboratory,Research,or Manufacturing Use
Normality (by titrimetry) Passes Test
(Makes 0.1N solution after dilution to 1000 mL)