Wipes for hand and surface cleaning, pre-saturated, Tork
Leverandør: SCA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Tork® Premium Tork®
115-0645EA 284 DKK
115-0645 115-0583
Wipes for hand and surface cleaning, pre-saturated, Tork
Papirservietter Engangsservietter og - håndklæder Imprægnerede servietter
Pre-moistened wipes made from a viscose and polyester mix. The single ply wipes are tough on grease and dirt but gentle enough on skin to use several times a day without risking dryness or irritation to skin.
  • Convenient, portable and recyclable dispensing container: Wipes are easy to dispense
  • Waterless cleaning: Tough on grease and dirt
  • Kind to skin and fragrance-free
Colour: White