VWR®, Exhaust Filters
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
590-1509EA 731 DKK
590-1509 590-1511
VWR®, Exhaust Filters
Filtre Ventilationsfiltre
These exhaust filters adsorb 99% of all volatile substances, vapours and gases, even organic solvents. They can be used on any HPLC system with multiple capillaries and connectors.
It is recommended that you exchange exhaust filter S every three months and the exhaust filter M every six months.