Inlet filters, low-pressure CPI, for HPLC system
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25009 25010
REST25009EA 938 DKK
REST25009 REST25010
Inlet filters, low-pressure CPI, for HPLC system
Filtre Væskebehandlingsfiltre
A 316 stainless steel knurled cap and Tefzel CPI ferrule seal to ¹/₈" Ext.Ø PTFE tubing when finger tightened onto the precision-machined filter holder.
  • Filter element is replaceable
  • Used as a helium sparging diffuser
  • 10 µm porosity protects delicate pump components