Tubing bender
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
REST23009EA 836 DKK
REST23009
Tubing bender
Tænger
These benders assists in making accurate left-hand, right-hand or offset bends. Bends 3,17 mm, 4,76 mm, or 6,35 mm tubing.