Y-PER™ Yeast Protein Extraction Reagent
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
78991 78990
PIER78991EA 1970 DKK
PIER78991 PIER78990
Y-PER™ Yeast Protein Extraction Reagent
Buffere Ekstraktions buffere
This reagent uses a mild detergent lysis procedure to efficiently release functionally active solubilised proteins.
  • Suitable for use in a diverse range of situations
  • Extracts more than twice as much protein as glass bead methods
  • Eliminates the physical problems associated with traditional glass bead lysis
  • Effective for many different organisms, including Gram-positive and Gram-negative bacteria
Y-PER reagent have been optimised to encompass a broad array of applications ranging from fusion-tagged protein purification to microplate-compatible enzyme assays, and genomic and plasmid DNA extraction from yeast. Also it has been used to isolate yeast killer virus double - stranded RNA from killer strains of Saccharomyces cerevisiae.