Luer access split septum device, Q-Syte™
Leverandør: BD Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BDAM385100EA 2510 DKK
BDAM385100 BDAM380510
Luer access split septum device, Q-Syte™
Kanyler
Split-septum device eliminating the complexities of mechanical valves and with them bacteria. Because of its straight and unobstructed path, it guarantees:
  • Higher flow rates and low priming volume
  • Smooth surface which is easily cleaned prior to access
  • Clear housing allows visual assessment of the fluid path
Specially designed for clinical applications.
Certificeringer: ISO 10555-5, Annex B.